missing translation for 'onlineSavingsMsg'
Learn More

2-Iodophenylacetic Acid 97.0+%, TCI America™

Catalog No. p-7131562
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
I07235G 5 g
I072325G 25 g
2 options

Catalog No. I07235G

Supplier: TCI America I07235G

Only null left

Chemical Identifiers

CAS 18698-96-9
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.046
MDL Number MFCD00046546
InChI Key IUHXGZHKSYYDIL-UHFFFAOYSA-N
Synonym 2-iodophenylacetic acid, 2-2-iodophenyl acetic acid, benzeneacetic acid, 2-iodo, 2-iodophenyl acetic acid, o-iodophenylacetic acid, 2-iodobenzeneacetic acid, 2-iodophenylaceticacid, pubchem2552, acmc-209eox, maybridge1_005605
PubChem CID 2780090
IUPAC Name 2-(2-iodophenyl)acetic acid
SMILES C1=CC=C(C(=C1)CC(=O)O)I

Specifications

Melting Point 117°C
Quantity 5 g
Formula Weight 262.05
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Iodophenylacetic Acid