missing translation for 'onlineSavingsMsg'
Learn More

2-Iodophenylacetic acid, 98+%, Thermo Scientific Chemicals

Catalog No. p-7046580
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1511606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1511606 Supplier Thermo Scientific Chemicals Supplier No. L1511606

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18698-96-9
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.046
MDL Number MFCD00046546
InChI Key IUHXGZHKSYYDIL-UHFFFAOYSA-N
Synonym 2-iodophenylacetic acid, 2-2-iodophenyl acetic acid, benzeneacetic acid, 2-iodo, 2-iodophenyl acetic acid, o-iodophenylacetic acid, 2-iodobenzeneacetic acid, 2-iodophenylaceticacid, pubchem2552, acmc-209eox, maybridge1_005605
PubChem CID 2780090
IUPAC Name 2-(2-iodophenyl)acetic acid
SMILES C1=CC=C(C(=C1)CC(=O)O)I

Specifications

Melting Point 115°C to 117°C
Quantity 5 g
Beilstein 2086800
Sensitivity Light sensitive
Formula Weight 262.05
Percent Purity ≥98%
Chemical Name or Material 2-Iodophenylacetic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only