missing translation for 'onlineSavingsMsg'
Learn More

2-Iodophenyl isothiocyanate, 97%, Thermo Scientific Chemicals

$99.22 - $99.22

Chemical Identifiers

CAS 98041-44-2
Molecular Formula C7H4INS
Molecular Weight (g/mol) 261.08
MDL Number MFCD00041057
InChI Key ODOTYLQYSDVPMA-UHFFFAOYSA-N
Synonym 2-iodophenyl isothiocyanate, 2-iodophenylisothiocyanate, o-iodophenyl isothiocyanate, 1-iodo-2-isothiocyanato-benzene, #, benzene,1-iodo-2-isothiocyanato, 1-iodanyl-2-isothiocyanato-benzene
PubChem CID 145310
IUPAC Name 1-iodo-2-isothiocyanatobenzene
SMILES IC1=CC=CC=C1N=C=S
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1101203
View Documents
Thermo Scientific Chemicals
L1101203
1 g
Each for $99.22
Only null left
 
Description

Description

2-Iodophenyl isothiocyanate is an important raw material and intermediate used in organic synthesis and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodophenyl isothiocyanate is an important raw material and intermediate used in organic synthesis and pharmaceuticals.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. It is sensitive to light and moisture. Incompatible with polyethers, polyester, alcohols, bases, amines and strong oxidizing agents.
Specifications

Chemical Identifiers

98041-44-2
261.08
ODOTYLQYSDVPMA-UHFFFAOYSA-N
145310
IC1=CC=CC=C1N=C=S
C7H4INS
MFCD00041057
2-iodophenyl isothiocyanate, 2-iodophenylisothiocyanate, o-iodophenyl isothiocyanate, 1-iodo-2-isothiocyanato-benzene, #, benzene,1-iodo-2-isothiocyanato, 1-iodanyl-2-isothiocyanato-benzene
1-iodo-2-isothiocyanatobenzene

Specifications

98041-44-2
>110°C (230°F)
MFCD00041057
UN2923
Light and moisture sensitive
ODOTYLQYSDVPMA-UHFFFAOYSA-N
1-iodo-2-isothiocyanatobenzene
145310
97%
36°C to 39°C
C7H4INS
1 g
2716369
2-iodophenyl isothiocyanate, 2-iodophenylisothiocyanate, o-iodophenyl isothiocyanate, 1-iodo-2-isothiocyanato-benzene, #, benzene,1-iodo-2-isothiocyanato, 1-iodanyl-2-isothiocyanato-benzene
IC1=CC=CC=C1N=C=S
261.08
261.09
2-Iodophenyl isothiocyanate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312+H332-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only