missing translation for 'onlineSavingsMsg'
Learn More

2-Iodophenyl isothiocyanate, 97%, Thermo Scientific Chemicals

Catalog No. AAL1101203
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1101203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1101203 Supplier Thermo Scientific Chemicals Supplier No. L1101203
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 98041-44-2 | Molecular Formula: C7H4INS | Molecular Weight: 261.08 g/mol

2-Iodophenyl isothiocyanate is an important raw material and intermediate used in organic synthesis and pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodophenyl isothiocyanate is an important raw material and intermediate used in organic synthesis and pharmaceuticals.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. It is sensitive to light and moisture. Incompatible with polyethers, polyester, alcohols, bases, amines and strong oxidizing agents.

Chemical Identifiers

CAS 98041-44-2
Molecular Formula C7H4INS
Molecular Weight (g/mol) 261.08
MDL Number MFCD00041057
InChI Key ODOTYLQYSDVPMA-UHFFFAOYSA-N
Synonym 2-iodophenyl isothiocyanate, 2-iodophenylisothiocyanate, o-iodophenyl isothiocyanate, 1-iodo-2-isothiocyanato-benzene, #, benzene,1-iodo-2-isothiocyanato, 1-iodanyl-2-isothiocyanato-benzene
PubChem CID 145310
IUPAC Name 1-iodo-2-isothiocyanatobenzene
SMILES IC1=CC=CC=C1N=C=S

Specifications

Melting Point 36°C to 39°C
Flash Point >110°C (230°F)
Quantity 1 g
UN Number UN2923
Beilstein 2716369
Sensitivity Light and moisture sensitive
Formula Weight 261.09
Percent Purity 97%
Chemical Name or Material 2-Iodophenyl isothiocyanate
Hazard Category H302+H312+H332-H314-H335
Hazard Statement GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only