missing translation for 'onlineSavingsMsg'
Learn More

2-(Iodomethyl)oxetane, 97%

Catalog No. p-7057145
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH66525MD 250 mg
AAH6652503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH66525MD Supplier Thermo Scientific Chemicals Supplier No. H66525MD
Only null left

CAS: 121138-00-9 | C4H7IO | 198.003 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 121138-00-9
Molecular Formula C4H7IO
Molecular Weight (g/mol) 198.003
MDL Number MFCD11226849
InChI Key JVPHNTROOBLPKR-UHFFFAOYSA-N
Synonym 2-iodomethyl oxetane, oxetane,2-iodomethyl
PubChem CID 49757971
IUPAC Name 2-(iodomethyl)oxetane
SMILES C1COC1CI

Specifications

Color Colorless to Yellow
Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 198
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-(Iodomethyl)oxetane
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only