accessibility menu, dialog, popup

UserName

OI

CAS RN 121138-00-9

IUPAC Name: 2-(iodomethyl)oxetane
Synonyms: 2-(iodomethyl)oxetane
Molecular Weight (g/mol): 198
Molecular Formula: C4H7IO
InChi Key: JVPHNTROOBLPKR-UHFFFAOYNA-N
SMILES: ICC1CCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results