missing translation for 'onlineSavingsMsg'
Learn More

2-Iodohippuric Acid 99.0+%, TCI America™

Catalog No. I04465G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04465G 5 g
I044625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I04465G Supplier TCI America Supplier No. I04465G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 147-58-0
Molecular Formula C9H8INO3
Molecular Weight (g/mol) 305.071
MDL Number MFCD00002694
InChI Key CORFWQGVBFFZHF-UHFFFAOYSA-N
Synonym N-(2-Iodobenzoyl)glycine
PubChem CID 8614
IUPAC Name 2-[(2-iodobenzoyl)amino]acetic acid
SMILES C1=CC=C(C(=C1)C(=O)NCC(=O)O)I

Specifications

Melting Point 173°C
Color White
Quantity 5 g
Formula Weight 305.07
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2-Iodohippuric Acid