missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Iodofluorene 95.0+%, TCI America™
Supplier: TCI America I07935G
Specifications
| 2-Iodofluorene | |
| 130°C | |
| C13H9I | |
| 5 g | |
| VNYQUOAQPXGXQO-UHFFFAOYSA-N | |
| 2-iodo-9H-fluorene | |
| 224094 | |
| ≥95.0% (GC) |
| 2523-42-4 | |
| White-Yellow | |
| MFCD00019049 | |
| 3077 | |
| C1C2=CC=CC=C2C3=C1C=C(C=C3)I | |
| 292.119 | |
| 292.12 | |
| Crystalline Powder |
Chemical Identifiers
| 2523-42-4 | |
| 292.119 | |
| VNYQUOAQPXGXQO-UHFFFAOYSA-N | |
| 2-iodo-9H-fluorene |
| C13H9I | |
| MFCD00019049 | |
| 224094 | |
| C1C2=CC=CC=C2C3=C1C=C(C=C3)I |
Safety and Handling
EINECSNumber : (4)-1782
TSCA : No