missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Iodobutane (stabilized with Copper chip) 98.0+%, TCI America™
$74.56 - $74.56
Chemical Identifiers
| CAS | 513-48-4 |
|---|---|
| Molecular Formula | C4H9I |
| Molecular Weight (g/mol) | 184.02 |
| MDL Number | MFCD00001072 |
| InChI Key | IQRUSQUYPCHEKN-UHFFFAOYNA-N |
| Synonym | sec-butyl iodide, butane, 2-iodo, 2-butyl iodide, sec-iodobutane, 2-jodbutan, 2-jodbutan czech, 2-iodanylbutane, 2-iodobutan, 2-iodo-butane, sec.-butyl iodide |
| PubChem CID | 10559 |
| IUPAC Name | 2-iodobutane |
| SMILES | CCC(C)I |
Chemical Identifiers
| 513-48-4 | |
| 184.02 | |
| IQRUSQUYPCHEKN-UHFFFAOYNA-N | |
| 10559 | |
| CCC(C)I |
| C4H9I | |
| MFCD00001072 | |
| sec-butyl iodide, butane, 2-iodo, 2-butyl iodide, sec-iodobutane, 2-jodbutan, 2-jodbutan czech, 2-iodanylbutane, 2-iodobutan, 2-iodo-butane, sec.-butyl iodide | |
| 2-iodobutane |
Specifications
| 513-48-4 | |
| C4H9I | |
| 25 g | |
| sec-butyl iodide, butane, 2-iodo, 2-butyl iodide, sec-iodobutane, 2-jodbutan, 2-jodbutan czech, 2-iodanylbutane, 2-iodobutan, 2-iodo-butane, sec.-butyl iodide | |
| CCC(C)I | |
| 184.02 | |
| 184.02 | |
| Liquid |
| 120°C | |
| MFCD00001072 | |
| 2390 | |
| IQRUSQUYPCHEKN-UHFFFAOYNA-N | |
| 2-iodobutane | |
| 10559 | |
| ≥98.0% (GC) | |
| 2-Iodobutane (stabilized with Copper chip) |
Safety and Handling
RTECSNumber : EK4410000
TSCA : Yes