missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Iodobutane, 99%, stab. with copper
Description
It is used as a solvent organic synthesis and as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a solvent organic synthesis and as pharmaceutical intermediate.
Solubility
Insoluble in water. Soluble in alcohol and diethyl ether.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
It is used as a solvent organic synthesis and as pharmaceutical intermediate.
Solubility
Insoluble in water. Soluble in alcohol and diethyl ether.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | -104°C |
| Density | 1.597 |
| Boiling Point | 119°C to 120°C |
| Flash Point | 21°C (70°F) |
| Odor | Ether-like |
| Refractive Index | 1.4991 |
| Linear Formula | CH3CH2CHICH3 |
| Quantity | 250 g |
| UN Number | UN2390 |
| Beilstein | 1718777 |
| Show More |
Safety and Handling
| Hazard Category | H226-H302+H332-H315-H319-H335 |
| Hazard Statement | GHS H Statement H225-H315-H319-H335 Highly flammable liquid and vapour. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 2-IODOBUTANE |
| EINECSNumber | 208-163-1 |
| RTECSNumber | EK4410000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only