missing translation for 'onlineSavingsMsg'
Learn More

2-Iodobutane, 99%, stab. with copper

Catalog No. AAB2061918
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2061918 50 g
AAB2061930 250 g
AAB2061936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2061918 Supplier Thermo Scientific Chemicals Supplier No. B2061918

CAS: 513-48-4 | C4H9I | 184.02 g/mol

It is used as a solvent organic synthesis and as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a solvent organic synthesis and as pharmaceutical intermediate.

Solubility
Insoluble in water. Soluble in alcohol and diethyl ether.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 513-48-4
Molecular Formula C4H9I
Molecular Weight (g/mol) 184.02
MDL Number MFCD00001072
InChI Key IQRUSQUYPCHEKN-UHFFFAOYNA-N
Synonym sec-butyl iodide, butane, 2-iodo, 2-butyl iodide, sec-iodobutane, 2-jodbutan, 2-jodbutan czech, 2-iodanylbutane, 2-iodobutan, 2-iodo-butane, sec.-butyl iodide
PubChem CID 10559
IUPAC Name 2-iodobutane
SMILES CCC(C)I

Specifications

Melting Point -104°C
Density 1.597
Boiling Point 119°C to 120°C
Flash Point 21°C (70°F)
Odor Ether-like
Refractive Index 1.4991
Linear Formula CH3CH2CHICH3
Quantity 50 g
UN Number UN2390
Beilstein 1718777
Sensitivity Light sensitive
Merck Index 14,1574
Solubility Information Insoluble in water. Soluble in alcohol and diethyl ether.
Formula Weight 184.02
Percent Purity 99%
Chemical Name or Material 2-Iodobutane, stabilized with copper
Show More Show Less
Hazard Category H226-H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 2-IODOBUTANE
EINECSNumber 208-163-1
RTECSNumber EK4410000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only