missing translation for 'onlineSavingsMsg'
Learn More

2-Iodobiphenyl 98.0+%, TCI America™

Catalog No. p-7131685
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I09675G 5 g
I096725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I09675G Supplier TCI America Supplier No. I09675G

Chemical Identifiers

CAS 2113-51-1
Molecular Formula C12H9I
Molecular Weight (g/mol) 280.108
MDL Number MFCD00039396
InChI Key QFUYDAGNUJWBSM-UHFFFAOYSA-N
Synonym 2-iodobiphenyl, 2-iodo-1,1'-biphenyl, o-iodobiphenyl, 1,1'-biphenyl, 2-iodo, o-phenyliodobenzene, 2-iodo-biphenyl, biphenyl, 2-iodo, 2-iodo-1-phenylbenzene, 1-iodo-2-phenyl-benzene, 1,1'-biphenyl, iodo
PubChem CID 75025
IUPAC Name 1-iodo-2-phenylbenzene
SMILES C1=CC=C(C=C1)C2=CC=CC=C2I

Specifications

Color Yellow
Boiling Point 158°C
Quantity 5 g
Formula Weight 280.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Iodobiphenyl
RTECSNumber DV5384800
TSCA Yes