missing translation for 'onlineSavingsMsg'
Learn More

2-Iodobiphenyl, 98%

Catalog No. AAL0699003
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0699003 1 g
AAL0699006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0699003 Supplier Thermo Scientific Chemicals Supplier No. L0699003

CAS: 2113-51-1 | C12H9I | 280.108 g/mol

2-Iodobiphenyl is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodobiphenyl is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2113-51-1
Molecular Formula C12H9I
Molecular Weight (g/mol) 280.108
MDL Number MFCD00039396
InChI Key QFUYDAGNUJWBSM-UHFFFAOYSA-N
Synonym 2-iodobiphenyl, 2-iodo-1,1'-biphenyl, o-iodobiphenyl, 1,1'-biphenyl, 2-iodo, o-phenyliodobenzene, 2-iodo-biphenyl, biphenyl, 2-iodo, 2-iodo-1-phenylbenzene, 1-iodo-2-phenyl-benzene, 1,1'-biphenyl, iodo
PubChem CID 75025
IUPAC Name 1-iodo-2-phenylbenzene
SMILES C1=CC=C(C=C1)C2=CC=CC=C2I

Specifications

Density 1.59
Boiling Point 300°C
Flash Point >110°C (230°F)
Refractive Index 1.661
Quantity 1 g
Beilstein 2206820
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 280.11
Percent Purity 98%
Chemical Name or Material 2-Iodobiphenyl
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-303-3
RTECSNumber DV5384800
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only