missing translation for 'onlineSavingsMsg'
Learn More

2-Iodobenzotrifluoride, 99%

Catalog No. p-7022785
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1569814 25 g
AAA1569822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1569814 Supplier Thermo Scientific Chemicals Supplier No. A1569814

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 444-29-1 | C7H4F3I | 272.009 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 444-29-1
Molecular Formula C7H4F3I
Molecular Weight (g/mol) 272.009
MDL Number MFCD00001041
InChI Key IGZGUYVVBABKOY-UHFFFAOYSA-N
Synonym 2-iodobenzotrifluoride, 1-iodo-2-trifluoromethyl benzene, o-iodobenzotrifluoride, benzene, 1-iodo-2-trifluoromethyl, 2-iodobenztrifluoride, o-trifluoromethyliodobenzene, 2-iodo-alpha,alpha,alpha-trifluorotoluene, iodobenzotrifluoride, 2-iodo-a,a,a-trifluorotoluene, 2-iodobenzotrifloride
PubChem CID 67957
IUPAC Name 1-iodo-2-(trifluoromethyl)benzene
SMILES C1=CC=C(C(=C1)C(F)(F)F)I

Specifications

Density 1.936
Boiling Point 198°C to 199°C
Flash Point 81°C (177°F)
Refractive Index 1.5305
Quantity 25 g
Beilstein 2090038
Sensitivity Light sensitive
Formula Weight 272.01
Percent Purity 99%
Chemical Name or Material 2-Iodobenzotrifluoride
Hazard Category H227-H314-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
EINECSNumber 207-147-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only