missing translation for 'onlineSavingsMsg'
Learn More

2-Iodobenzonitrile 98.0+%, TCI America™

Catalog No. I065925G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I065925G 25 g
I06591G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I065925G Supplier TCI America Supplier No. I065925G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 4387-36-4
Molecular Formula C7H4IN
Molecular Weight (g/mol) 229.02
InChI Key JDDAFHUEOVUDFJ-UHFFFAOYSA-N
Synonym 1-cyano-2-iodobenzene, benzonitrile, 2-iodo, o-iodobenzonitrile, 2-iodobenzenecarbonitrile, iodobenzonitrile, 2-cyanoiodobenzene, pubchem3932, acmc-209jvq, 2-iodobenzonitrile, ksc235m8d
PubChem CID 2759358
IUPAC Name 2-iodobenzonitrile
SMILES C1=CC=C(C(=C1)C#N)I

Specifications

Melting Point 55°C
Boiling Point 147°C
Quantity 25 g
UN Number 3439
Formula Weight 229.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Iodobenzonitrile