missing translation for 'onlineSavingsMsg'
Learn More

2-Iodobenzamide 98.0+%, TCI America™

Catalog No. p-7131569
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07345G 5 g
I073425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07345G Supplier TCI America Supplier No. I07345G

Chemical Identifiers

CAS 3930-83-4
Molecular Formula C7H6INO
Molecular Weight (g/mol) 247.035
MDL Number MFCD00099248
InChI Key YEOYYWCXWUDVCX-UHFFFAOYSA-N
PubChem CID 77529
IUPAC Name 2-iodobenzamide
SMILES C1=CC=C(C(=C1)C(=O)N)I

Specifications

Melting Point 184°C
Quantity 5 g
Formula Weight 247.04
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2-Iodobenzamide
RTECSNumber CV5399625
TSCA No