missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Iodobenzamide 98.0+%, TCI America™
Supplier: TCI America I073425G
Specifications
| 2-Iodobenzamide | |
| 184°C | |
| MFCD00099248 | |
| YEOYYWCXWUDVCX-UHFFFAOYSA-N | |
| 2-iodobenzamide | |
| 77529 | |
| ≥98.0% (GC,N) |
| 3930-83-4 | |
| C7H6INO | |
| 25 g | |
| C1=CC=C(C(=C1)C(=O)N)I | |
| 247.035 | |
| 247.04 | |
| Crystalline Powder |
Chemical Identifiers
| 3930-83-4 | |
| 247.035 | |
| YEOYYWCXWUDVCX-UHFFFAOYSA-N | |
| 2-iodobenzamide |
| C7H6INO | |
| MFCD00099248 | |
| 77529 | |
| C1=CC=C(C(=C1)C(=O)N)I |
Safety and Handling
RTECSNumber : CV5399625
TSCA : No