missing translation for 'onlineSavingsMsg'
Learn More

2-Iodoanisole (stabilized with Copper chip) 98.0+%, TCI America™

Catalog No. I038225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I038225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I038225G Supplier TCI America Supplier No. I038225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 529-28-2
Molecular Formula C7H7IO
Molecular Weight (g/mol) 234.036
MDL Number MFCD00001039
InChI Key DVQWNQBEUKXONL-UHFFFAOYSA-N
Synonym 2-iodoanisole, o-iodoanisole, benzene, 1-iodo-2-methoxy, iodoanisole, 2-methoxyiodobenzene, o-anisyl iodide, 2-iodophenol methyl ether, anisole, o-iodo, 2-iodophenyl methyl ether, 1-iodo-2-methoxy-benzene
PubChem CID 68257
ChEBI CHEBI:16355
IUPAC Name 1-iodo-2-methoxybenzene
SMILES COC1=CC=CC=C1I

Specifications

Color Yellow
Boiling Point 238°C
Quantity 25 g
Formula Weight 234.04
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Iodoanisole (stabilized with Copper chip)