missing translation for 'onlineSavingsMsg'
Learn More

2-Iodoanisole, 99%

Catalog No. p-7080569
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1015014 25 g
AAA1015022 100 g
AAA1015036 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1015014 Supplier Thermo Scientific Chemicals Supplier No. A1015014

CAS: 529-28-2 | C7H7IO | 234.036 g/mol

2-Iodoanisole has been used in palladium/copper-catalyzed synthesis of o-(1-alkynyl)anisoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodoanisole has been used in palladium/copper-catalyzed synthesis of o-(1-alkynyl)anisoles.

Solubility
Miscible with alcohol, chloroform and diethyl ether. Insoluble in water.

Notes
Light Sensitive, store in dark. Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.

Chemical Identifiers

CAS 529-28-2
Molecular Formula C7H7IO
Molecular Weight (g/mol) 234.036
MDL Number MFCD00001039
InChI Key DVQWNQBEUKXONL-UHFFFAOYSA-N
Synonym 2-iodoanisole, o-iodoanisole, benzene, 1-iodo-2-methoxy, iodoanisole, 2-methoxyiodobenzene, o-anisyl iodide, 2-iodophenol methyl ether, anisole, o-iodo, 2-iodophenyl methyl ether, 1-iodo-2-methoxy-benzene
PubChem CID 68257
ChEBI CHEBI:16355
IUPAC Name 1-iodo-2-methoxybenzene
SMILES COC1=CC=CC=C1I

Specifications

Density 1.8
Boiling Point 240°C to 242°C
Flash Point >110°C (230°F)
Refractive Index 1.622
Quantity 25 g
Beilstein 1860243
Sensitivity Light sensitive
Merck Index 14,5028
Solubility Information Miscible with alcohol,chloroform and diethyl ether. Insoluble in water.
Formula Weight 234.04
Percent Purity 99%
Chemical Name or Material 2-Iodoanisole
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 208-456-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only