missing translation for 'onlineSavingsMsg'
Learn More

2'-Iodoacetophenone, 99+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 393130010

 View more versions of this product

Catalog No. AC393130010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2'-Iodoacetophenone
99.0
1.7460g/mL
Authentic
Glass bottle
1.617 to 1.619 (20°C, 589nm)
MFCD00094998
1.746
Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
CC(=O)C1=CC=CC=C1I
246.04
246.04
2142-70-3
100.0
139°C to 140°C (12.0mmHg)
99% min. (GC)
C8H7IO
IC6H4C(O)CH3
1g
2'-iodoacetophenone, 1-2-iodophenyl ethanone, o-iodoacetophenone, 1-2-iodophenyl ethan-1-one, o-iodobenzoyl, acetophenone, o-iodo, acetophenone, 2'-iodo, ethanone, 1-iodophenyl, ethanone, 1-2-iodophenyl, 1-acetyl-2-iodobenzene
XDXCBCXNCQGZPG-UHFFFAOYSA-N
1-(2-iodophenyl)ethanone
240431
99+%

Chemical Identifiers

2142-70-3
246.04
XDXCBCXNCQGZPG-UHFFFAOYSA-N
240431
CC(=O)C1=CC=CC=C1I
C8H7IO
MFCD00094998
2'-iodoacetophenone, 1-2-iodophenyl ethanone, o-iodoacetophenone, 1-2-iodophenyl ethan-1-one, o-iodobenzoyl, acetophenone, o-iodo, acetophenone, 2'-iodo, ethanone, 1-iodophenyl, ethanone, 1-2-iodophenyl, 1-acetyl-2-iodobenzene
1-(2-iodophenyl)ethanone
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only