missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-5-(trifluoromethyl)pyridine, 96%

Catalog No. AAH6384803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6384803 1 g
AAH6384806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6384803 Supplier Thermo Scientific Chemicals Supplier No. H6384803
Only null left

CAS: 100366-75-4 | C6H3F3IN | 273.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 100366-75-4
Molecular Formula C6H3F3IN
Molecular Weight (g/mol) 273.00
MDL Number MFCD08741351
InChI Key WSEVWIIOEQZEOU-UHFFFAOYSA-N
PubChem CID 22147841
IUPAC Name 2-iodo-5-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=CN=C(I)C=C1

Specifications

Quantity 1 g
Sensitivity Light sensitive
Synonym 2-iodo-5-trifluoromethyl pyridine, 5-trifluoromethyl-2-iodopyridine, 2-iodo-5-trifluoromethyl-pyridine, pyridine, 2-iodo-5-trifluoromethyl, pubchem16308, acmc-1c5wh, 2-iodanyl-5-trifluoromethyl pyridine, pyridine,2-iodo-5-trifluoromethyl
Formula Weight 272.99
Percent Purity 96%
Chemical Name or Material 2-Iodo-5-(trifluoromethyl)pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only