accessibility menu, dialog, popup

UserName

CAS RN 100366-75-4

IUPAC Name: 2-iodo-5-(trifluoromethyl)pyridine
Synonyms: 2-iodo-5-(trifluoromethyl)pyridine
Molecular Weight (g/mol): 273
Molecular Formula: C6H3F3IN
InChi Key: WSEVWIIOEQZEOU-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CN=C(I)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results