missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Iodo-5-methylaniline 98.0+%, TCI America™
Supplier: TCI America I10341G
Specifications
| 2-Iodo-5-methylaniline | |
| 36°C | |
| MFCD00833395 | |
| 3-Amino-4-iodotoluene | |
| CC1=CC(=C(C=C1)I)N | |
| 233.052 | |
| 233.05 | |
| Crystalline Powder |
| 13194-69-9 | |
| C7H8IN | |
| 1 g | |
| KXPBTNCFONSVIA-UHFFFAOYSA-N | |
| 2-iodo-5-methylaniline | |
| 2736659 | |
| ≥98.0% (GC,T) |
Chemical Identifiers
| 13194-69-9 | |
| 233.052 | |
| KXPBTNCFONSVIA-UHFFFAOYSA-N | |
| 2736659 | |
| CC1=CC(=C(C=C1)I)N |
| C7H8IN | |
| MFCD00833395 | |
| 3-Amino-4-iodotoluene | |
| 2-iodo-5-methylaniline |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator