missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-5-methoxybenzaldehyde, 96%

Catalog No. AAH6616303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6616303 1 g
AAH6616306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6616303 Supplier Thermo Scientific Chemicals Supplier No. H6616303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 77287-58-2 | C8H7IO2 | 262.046 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 77287-58-2
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.046
MDL Number MFCD18905098
InChI Key YHRFMOFYYPMQBE-UHFFFAOYSA-N
Synonym benzaldehyde, 2-iodo-5-methoxy, 5-methoxy-2-iodobenzaldehyde, 2-iodo-5-methoxybenzaldehyde
PubChem CID 10858258
IUPAC Name 2-iodo-5-methoxybenzaldehyde
SMILES COC1=CC(=C(C=C1)I)C=O

Specifications

Melting Point 110°C to 115°C
Color White to Yellow
Quantity 1 g
Sensitivity Air and light sensitive
Formula Weight 262.04
Percent Purity 96%
Physical Form Solid
Chemical Name or Material 2-Iodo-5-methoxybenzaldehyde
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only