missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-4-(trifluoromethyl)aniline, 99%

Catalog No. p-7026609
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2269109 10 g
AAB2269118 50 g
AAB2269130 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2269109 Supplier Thermo Scientific Chemicals Supplier No. B2269109

CAS: 163444-17-5 | C7H5F3IN | 287.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 163444-17-5
Molecular Formula C7H5F3IN
Molecular Weight (g/mol) 287.02
MDL Number MFCD02683097
InChI Key UKKWTZPXYIYONW-UHFFFAOYSA-N
Synonym 4-amino-3-iodobenzotrifluoride, 2-iodo-4-trifluoromethyl aniline, 4-trifluoromethyl-2-iodobenzenamine, benzenamine, 2-iodo-4-trifluoromethyl, 2-iodo-4-trifluoromethyl benzenamine, pubchem1632, acmc-209dpp, intermediates-zcf02228, ksc910s3b, 3-iodo-4-aminobenzotrifluoride
PubChem CID 2783322
IUPAC Name 2-iodo-4-(trifluoromethyl)aniline
SMILES NC1=C(I)C=C(C=C1)C(F)(F)F

Specifications

Melting Point 49°C to 52°C
Quantity 10 g
Sensitivity Light sensitive
Formula Weight 287.02
Percent Purity 99%
Chemical Name or Material 2-Iodo-4-(trifluoromethyl)aniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only