missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-3-nitrotoluene 98.0+%, TCI America™

Catalog No. I07995G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I07995G 5 g
I079925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I07995G Supplier TCI America Supplier No. I07995G
Only null left

Chemical Identifiers

CAS 6277-17-4
Molecular Formula C7H6INO2
Molecular Weight (g/mol) 263.034
MDL Number MFCD00091040
InChI Key AKYPSJARSQSCHJ-UHFFFAOYSA-N
Synonym 2-Iodo-1-methyl-3-nitrobenzene
PubChem CID 234781
IUPAC Name 2-iodo-1-methyl-3-nitrobenzene
SMILES CC1=C(C(=CC=C1)[N+](=O)[O-])I

Specifications

Melting Point 67°C
Color Yellow
Quantity 5 g
Formula Weight 263.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Iodo-3-nitrotoluene