missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-3-methylbenzoic acid, 98%

Catalog No. p-7047877
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1987603 1 g
AAL1987606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1987603 Supplier Thermo Scientific Chemicals Supplier No. L1987603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 108078-14-4 | C8H7IO2 | 262.046 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 108078-14-4
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.046
MDL Number MFCD00079764
InChI Key PZUXUOZSSYKAMX-UHFFFAOYSA-N
Synonym 2-iodo-m-toluic acid, benzoic acid,2-iodo-3-methyl, 2-iodo-3-methyl-benzoic acid, pubchem12741, acmc-1bsuz, 3-methyl-2-iodobenzoic acid, 2-iodo-3-methyl benzoic acid, buttpark 148\07-11, 2-iodo-3-methylbenzoic acid
PubChem CID 2759365
IUPAC Name 2-iodo-3-methylbenzoic acid
SMILES CC1=CC=CC(=C1I)C(=O)O

Specifications

Melting Point 150°C to 153°C
Quantity 1 g
Beilstein 2439521
Sensitivity Light sensitive
Formula Weight 262.05
Percent Purity 98%
Chemical Name or Material 2-Iodo-3-methylbenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only