missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-1,3,5-trimethylbenzene, 98%

Catalog No. AAA1536006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1536006 5 g
AAA1536014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1536006 Supplier Thermo Scientific Chemicals Supplier No. A1536006
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 4028-63-1 | C9H11I | 246.091 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4028-63-1
Molecular Formula C9H11I
Molecular Weight (g/mol) 246.091
MDL Number MFCD00013707
InChI Key GTPNXFKONRIHRW-UHFFFAOYSA-N
Synonym 2,4,6-trimethyliodobenzene, 2-iodomesitylene, mesityl iodide, benzene, 2-iodo-1,3,5-trimethyl, iodo-2,4,6-trimethylbenzene, 2,4,6-trimethyliodobenzene 2-iodomesitylene, iodomesitylene, jodmesitylen, mesityliodine, pubchem12601
PubChem CID 77647
IUPAC Name 2-iodo-1,3,5-trimethylbenzene
SMILES CC1=CC(=C(C(=C1)C)I)C

Specifications

Melting Point 28°C to 30°C
Quantity 5 g
Beilstein 1860064
Sensitivity Light sensitive
Formula Weight 246.09
Percent Purity 98%
Chemical Name or Material 2-Iodo-1,3,5-trimethylbenzene
EINECSNumber 223-709-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only