missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| 2-Indanone | |
| 57°C | |
| C9H8O | |
| 25 g | |
| UMJJFEIKYGFCAT-UHFFFAOYSA-N | |
| 1,3-dihydroinden-2-one | |
| 11983 | |
| 132.16 | |
| Crystalline Powder |
| 615-13-4 | |
| 225°C | |
| MFCD00003792 | |
| 2-indanone, indan-2-one, 1h-inden-2 3h-one, 1,3-dihydro-2h-inden-2-one, beta-hydrindone, 2h-inden-2-one, 1,3-dihydro, unii-0i79n673de, 2,3-dihydro-1h-inden-2-one, 4-07-00-01002 beilstein handbook reference, 2-lndanone | |
| C1C(=O)CC2=CC=CC=C21 | |
| 132.162 | |
| CHEBI:27930 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 615-13-4 | |
| 132.162 | |
| UMJJFEIKYGFCAT-UHFFFAOYSA-N | |
| 11983 | |
| 1,3-dihydroinden-2-one |
| C9H8O | |
| MFCD00003792 | |
| 2-indanone, indan-2-one, 1h-inden-2 3h-one, 1,3-dihydro-2h-inden-2-one, beta-hydrindone, 2h-inden-2-one, 1,3-dihydro, unii-0i79n673de, 2,3-dihydro-1h-inden-2-one, 4-07-00-01002 beilstein handbook reference, 2-lndanone | |
| CHEBI:27930 | |
| C1C(=O)CC2=CC=CC=C21 |
Safety and Handling
RTECSNumber : NK7535500
TSCA : Yes
Recommended Storage : Refrigerator