missing translation for 'onlineSavingsMsg'
Learn More

2-Indanone 98.0+%, TCI America™

Catalog No. p-7131523
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
I06495G 5 g
I064925G 25 g
2 options

Catalog No. I06495G

Supplier: TCI America I06495G

Only null left

Chemical Identifiers

CAS 615-13-4
Molecular Formula C9H8O
Molecular Weight (g/mol) 132.162
MDL Number MFCD00003792
InChI Key UMJJFEIKYGFCAT-UHFFFAOYSA-N
Synonym 2-indanone, indan-2-one, 1h-inden-2 3h-one, 1,3-dihydro-2h-inden-2-one, beta-hydrindone, 2h-inden-2-one, 1,3-dihydro, unii-0i79n673de, 2,3-dihydro-1h-inden-2-one, 4-07-00-01002 beilstein handbook reference, 2-lndanone
PubChem CID 11983
ChEBI CHEBI:27930
IUPAC Name 1,3-dihydroinden-2-one
SMILES C1C(=O)CC2=CC=CC=C21

Specifications

Melting Point 57°C
Boiling Point 225°C
Quantity 5 g
Formula Weight 132.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Indanone
RTECSNumber NK7535500
TSCA Yes
Recommended Storage Refrigerator