Learn More
2-Imidazolidone, 96%, anhydrous
CAS: 120-93-4 | C3H6N2O | 86.09 g/mol
$182.32 - $182.32
Chemical Identifiers
| CAS | 120-93-4 |
|---|---|
| Molecular Formula | C3H6N2O |
| Molecular Weight (g/mol) | 86.09 |
| MDL Number | MFCD00005257 |
| InChI Key | YAMHXTCMCPHKLN-UHFFFAOYSA-N |
| Synonym | 2-imidazolidone, 2-imidazolidinone, ethyleneurea, imidazolidinone, 2-oxoimidazolidine, ethylene urea, n,n'-ethyleneurea, 1,3-ethyleneurea, imidazolid-2-one, monoethyleneurea |
| PubChem CID | 8453 |
| ChEBI | CHEBI:37001 |
| IUPAC Name | imidazolidin-2-one |
| SMILES | O=C1NCCN1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC412272500
|
Thermo Scientific Chemicals
412272500 |
250 g | Glass bottle |
Each for $182.32
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 120-93-4 | |
| 86.09 | |
| YAMHXTCMCPHKLN-UHFFFAOYSA-N | |
| 8453 | |
| imidazolidin-2-one |
| C3H6N2O | |
| MFCD00005257 | |
| 2-imidazolidone, 2-imidazolidinone, ethyleneurea, imidazolidinone, 2-oxoimidazolidine, ethylene urea, n,n'-ethyleneurea, 1,3-ethyleneurea, imidazolid-2-one, monoethyleneurea | |
| CHEBI:37001 | |
| O=C1NCCN1 |
Specifications
| 120-93-4 | |
| 100.0 | |
| White | |
| Authentic | |
| Glass bottle | |
| MFCD00005257 | |
| 24,2 | |
| 2-imidazolidone, 2-imidazolidinone, ethyleneurea, imidazolidinone, 2-oxoimidazolidine, ethylene urea, n,n'-ethyleneurea, 1,3-ethyleneurea, imidazolid-2-one, monoethyleneurea | |
| O=C1NCCN1 | |
| 86.09 | |
| CHEBI:37001 | |
| 96% | |
| 2-Imidazolidone |
| 95.0 | |
| 131.4°C to 132.4°C | |
| 187.0°C (10.0 Torr) | |
| 95% min. (HPLC) | |
| C3H6N2O | |
| 250 g | |
| 15,4955 | |
| YAMHXTCMCPHKLN-UHFFFAOYSA-N | |
| imidazolidin-2-one | |
| 8453 | |
| 86.09 | |
| Crystalline Powder or Powder |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 204-436-4
RUO – Research Use Only