missing translation for 'onlineSavingsMsg'
Learn More

2-Imidazolidone, 96%, anhydrous

Catalog No. AC412272500
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC412272500 250 g Glass bottle
AC412270250 25 g Glass bottle
AC412270010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC412272500 Supplier Thermo Scientific Chemicals Supplier No. 412272500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 120-93-4 | C3H6N2O | 86.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 120-93-4
Molecular Formula C3H6N2O
Molecular Weight (g/mol) 86.09
MDL Number MFCD00005257
InChI Key YAMHXTCMCPHKLN-UHFFFAOYSA-N
Synonym 2-imidazolidone, 2-imidazolidinone, ethyleneurea, imidazolidinone, 2-oxoimidazolidine, ethylene urea, n,n'-ethyleneurea, 1,3-ethyleneurea, imidazolid-2-one, monoethyleneurea
PubChem CID 8453
ChEBI CHEBI:37001
IUPAC Name imidazolidin-2-one
SMILES O=C1NCCN1

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 131.4°C to 132.4°C
Color White
Boiling Point 187.0°C (10.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 250 g
Beilstein 24,2
Merck Index 15,4955
Formula Weight 86.09
Percent Purity 96%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 2-Imidazolidone
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 204-436-4

RUO – Research Use Only