missing translation for 'onlineSavingsMsg'
Learn More

2-Imidazolidinone 97.0+%, TCI America™

Catalog No. I000525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I000525G 25 g
I0005500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I000525G Supplier TCI America Supplier No. I000525G

Chemical Identifiers

CAS 120-93-4
Molecular Formula C3H6N2O
Molecular Weight (g/mol) 86.09
MDL Number MFCD00005257
InChI Key YAMHXTCMCPHKLN-UHFFFAOYSA-N
Synonym 2-imidazolidone, 2-imidazolidinone, ethyleneurea, imidazolidinone, 2-oxoimidazolidine, ethylene urea, n,n'-ethyleneurea, 1,3-ethyleneurea, imidazolid-2-one, monoethyleneurea
PubChem CID 8453
ChEBI CHEBI:37001
IUPAC Name imidazolidin-2-one
SMILES O=C1NCCN1

Specifications

Melting Point 132°C
Color White-Yellow
pH 10.5 (100g/L H2O 20°C)
Quantity 25 g
Formula Weight 86.09
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-Imidazolidinone
EINECSNumber (2)-1736&(5)-0431
RTECSNumber NJ0570000
TSCA Yes