missing translation for 'onlineSavingsMsg'
Learn More

2-Imidazolidinone hemihydrate, 98+%

Catalog No. AAA1766630
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1766630 250 g
AAA176660B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1766630 Supplier Thermo Scientific Chemicals Supplier No. A1766630
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 121325-67-5 | C6H14N4O3 | 190.20 g/mol

2-Imidazolidinone hemihydrate is used as a scavenger for formaldehyde. It is utilized for removal of formaldehyde from leather, rubber and textile coating. It is also used as a raw material for the preparation of active pharmaceutical ingredients like azlocillin sodium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Imidazolidinone hemihydrate is used as a scavenger for formaldehyde. It is utilized for removal of formaldehyde from leather, rubber and textile coating. It is also used as a raw material for the preparation of active pharmaceutical ingredients like azlocillin sodium.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 121325-67-5
Molecular Formula C6H14N4O3
Molecular Weight (g/mol) 190.20
MDL Number MFCD02662355
InChI Key KPRJGGOOWATRNT-UHFFFAOYSA-N
Synonym imidazolidin-2-one hydrate, 2-imidazolidinone hemihydrate, 2-imidazolidone hemihydrate, bis imidazolidin-2-one hydrate, bis imidazolidinone hydrate, 2-imidazolidinone, hydrate 2:1, imidazolidin-2-one, imidazolidin-2-one, hydrate
PubChem CID 2723651
IUPAC Name imidazolidin-2-one;hydrate
SMILES O.O=C1NCCN1.O=C1NCCN1

Specifications

Melting Point 55°C
Quantity 250 g
Solubility Information Soluble in water.
Formula Weight 95.10 (86.09 Anhydrous)
Percent Purity ≥98%
Chemical Name or Material 2-Imidazolidinone hemihydrate
Hazard Category H319-H361f
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only