Learn More
2-Imidazolidinethione, 98%
CAS: 96-45-7 | C3H6N2S | 102.16 g/mol
$125.37 - $125.37
Chemical Identifiers
| CAS | 96-45-7 |
|---|---|
| Molecular Formula | C3H6N2S |
| Molecular Weight (g/mol) | 102.16 |
| MDL Number | MFCD00005276 |
| InChI Key | PDQAZBWRQCGBEV-UHFFFAOYSA-N |
| Synonym | 2-imidazolidinethione, ethylenethiourea, ethylene thiourea, 2-mercaptoimidazoline, imidazolidinethione, n,n'-ethylenethiourea, mercazin i, warecure c, rhenogran etu, mercaptoimidazoline |
| PubChem CID | 2723650 |
| ChEBI | CHEBI:34750 |
| IUPAC Name | imidazolidine-2-thione |
| SMILES | S=C1NCCN1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC122032500
|
Thermo Scientific Chemicals
122032500 |
250 g | Glass bottle |
N/A
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 96-45-7 | |
| 102.16 | |
| PDQAZBWRQCGBEV-UHFFFAOYSA-N | |
| 2723650 | |
| imidazolidine-2-thione |
| C3H6N2S | |
| MFCD00005276 | |
| 2-imidazolidinethione, ethylenethiourea, ethylene thiourea, 2-mercaptoimidazoline, imidazolidinethione, n,n'-ethylenethiourea, mercazin i, warecure c, rhenogran etu, mercaptoimidazoline | |
| CHEBI:34750 | |
| S=C1NCCN1 |
Specifications
| 96-45-7 | |
| 100.0 | |
| White | |
| Authentic | |
| Glass bottle | |
| MFCD00005276 | |
| 24, 4 | |
| 2-imidazolidinethione, ethylenethiourea, ethylene thiourea, 2-mercaptoimidazoline, imidazolidinethione, n,n'-ethylenethiourea, mercazin i, warecure c, rhenogran etu, mercaptoimidazoline | |
| PDQAZBWRQCGBEV-UHFFFAOYSA-N | |
| imidazolidine-2-thione | |
| 2723650 | |
| 102.15 | |
| Powder |
| 97.5 | |
| 197.0°C to 201.0°C | |
| 252°C | |
| 97.5% min. (HPLC) | |
| C3H6N2S | |
| 250 g | |
| 15, 3858 | |
| Solubility in water: 19g/L (20°C). Other solubilities: 440g/L water (90°C), moderately soluble in methanol, ethanol, ethylene, glycol and pyridine, insoluble in acetone, ether, chloroform, benzene, and ligroin | |
| S=C1NCCN1 | |
| 102.16 | |
| CHEBI:34750 | |
| 98% | |
| 2-Imidazolidinethione, 98% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Suspected of causing cancer.
May damage the unborn child.
Causes damage to organs through prolonged or repeated exposure if inhaled.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
GHS Signal Word: Danger
EINECSNumber : 202-506-9
RTECSNumber : NI9625000
TSCA : TSCA
RUO – Research Use Only