missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxypyridine N-Oxide 98.0+%, TCI America™

Catalog No. H067225G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H067225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H067225G Supplier TCI America Supplier No. H067225G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Superior, non-explosive coupling additive used to enhance the synthesis of active pharmaceutical ingredients and suppress racemization.

Chemical Identifiers

CAS 13161-30-3
Molecular Formula C5H5NO2
Molecular Weight (g/mol) 111.1
MDL Number MFCD00006195
InChI Key SNUSZUYTMHKCPM-UHFFFAOYSA-N
Synonym 2-pyridinol-1-oxide, 2-hydroxypyridine 1-oxide, 2-hydroxypyridine n-oxide, 1-hydroxypyridin-2 1h-one, 2-hydroxypyridine-n-oxide, hydroxypyridinone, pyridinol, 1-oxide, 2-pyridinol 1-oxide, 1-hydroxy-2-pyridone, 1-hydroxy-1,2-dihydropyridin-2-one
PubChem CID 69975
IUPAC Name 1-hydroxypyridin-2-one
SMILES C1=CC(=O)N(C=C1)O

Specifications

Melting Point 151°C
Quantity 25 g
Formula Weight 111.10
Percent Purity ≥98.0% (T)
Grade Reagent
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxypyridine N-Oxide