accessibility menu, dialog, popup

UserName

OHNO

CAS RN 13161-30-3

IUPAC Name: 1-hydroxy-1,2-dihydropyridin-2-one
Synonyms: 1-hydroxypyridin-2-one
Molecular Weight (g/mol): 111.1
Molecular Formula: C5H5NO2
InChi Key: SNUSZUYTMHKCPM-UHFFFAOYSA-N
SMILES: ON1C=CC=CC1=O
Percent Purity 
  • (3)
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (6)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)