missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxypyridine-N-oxide, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 243791000

Quantity 100g

 View more versions of this product

Catalog No. AC243791000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

13161-30-3
111.1
2-pyridinol-1-oxide, 2-hydroxypyridine 1-oxide, 2-hydroxypyridine n-oxide, 1-hydroxypyridin-2 1h-one, 2-hydroxypyridine-n-oxide, hydroxypyridinone, pyridinol, 1-oxide, 2-pyridinol 1-oxide, 1-hydroxy-2-pyridone, 1-hydroxy-1,2-dihydropyridin-2-one
1-hydroxypyridin-2-one
C5H5NO2
SNUSZUYTMHKCPM-UHFFFAOYSA-N
69975
C1=CC(=O)N(C=C1)O

Specifications

2-Hydroxypyridine-N-oxide
100°C
C5H5NO2
2-pyridinol-1-oxide, 2-hydroxypyridine 1-oxide, 2-hydroxypyridine n-oxide, 1-hydroxypyridin-2 1h-one, 2-hydroxypyridine-n-oxide, hydroxypyridinone, pyridinol, 1-oxide, 2-pyridinol 1-oxide, 1-hydroxy-2-pyridone, 1-hydroxy-1,2-dihydropyridin-2-one
C1=CC(=O)N(C=C1)O
111.1
111.1
13161-30-3
Authentic
100g
SNUSZUYTMHKCPM-UHFFFAOYSA-N
1-hydroxypyridin-2-one
69975
97%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 236-100-8

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...