Learn More
(2-Hydroxypropyl)-beta-cyclodextrin, MW ca 1250-1480
CAS: 128446-35-5 | C44H75O36 | 1180.05 g/mol
Supplier: Thermo Scientific Chemicals H3113314
Description
2-Hydroxypropyl)-β-cyclodextrin is used to enhance solubility of hydrophobic compounds by forming complex with them. This favors its usage in pharmaceutics, in the manufacturing of drugs. Beta- cyclodextrin and its methyl derivative is used to remove cholesterol from cultured cells. Many common chiral stationary phases used in high-performance liquid chromatography columns consists of beta-cyclodextrin.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Hydroxypropyl)-β-cyclodextrin is used to enhance solubility of hydrophobic compounds by forming complex with them. This favors its usage in pharmaceutics, in the manufacturing of drugs. Beta- cyclodextrin and its methyl derivative is used to remove cholesterol from cultured cells. Many common chiral stationary phases used in high-performance liquid chromatography columns consists of beta-cyclodextrin.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents. Keep the container tightly closed in a dry and well-ventilated place.
Specifications
| (2-Hydroxypropyl)-beta-cyclodextrin | |
| 305°C | |
| MFCD00069372 | |
| Soluble in water. | |
| ≈1250 to 1480 d |
| 128446-35-5 | |
| C44H75O36 | |
| 25 g | |
| 1180.05 |
Chemical Identifiers
| 128446-35-5 | |
| C44H75O36 | |
| 1180.05 | |
| MFCD00069372 |
Safety and Handling
EINECSNumber : 420-920-1
RTECSNumber : GU2293332
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only