missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. H11841G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H11841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H11841G Supplier TCI America Supplier No. H11841G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 2-Hydroxyphenylboronic Acid (contains varying amounts of Anhydride)
CAS 89466-08-0
Melting Point 185°C
Molecular Formula C6H7BO3
Quantity 1 g
MDL Number MFCD01074581
Synonym 2-hydroxyphenyl boronic acid, 2-hydroxybenzeneboronic acid, o-hydroxyphenylboronic acid, 2-boronophenol, 2-hydroxyphenyl boranediol, 2-hydroxyphenylboronicacid, boronic acid, 2-hydroxyphenyl, pubchem9522, 2-hydroxyphenylboronic, acmc-209r1d
InChI Key YDMRDHQUQIVWBE-UHFFFAOYSA-N
SMILES OB(O)C1=CC=CC=C1O
IUPAC Name (2-hydroxyphenyl)boronic acid
Molecular Weight (g/mol) 137.93
PubChem CID 2773454
Formula Weight 137.93
Physical Form Crystalline Powder
Show More Show Less