missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyphenoxyacetic acid, 98+%

Catalog No. AAL0389706
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0389706 5 g
AAL0389714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0389706 Supplier Thermo Scientific Chemicals Supplier No. L0389706
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6324-11-4 | C8H8O4 | 168.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6324-11-4
MDL Number MFCD00015454

Specifications

Melting Point 125°C to 130°C
Molecular Formula C8H8O4
Quantity 5 g
Beilstein 1948566
Synonym 2-hydroxyphenoxyacetic acid, 2-hydroxyphenoxy acetic acid, 2-2-hydroxyphenoxy acetic acid, o-hydroxyphenoxy acetic acid, acetic acid, hydroxyphenoxy, o-hydroxyphenoxyacetic acid, enamine_004711, 2-carboxymethoxy phenol, cambridge id 5156176
InChI Key LPXHPAYRONCQIF-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)O)OCC(=O)O
IUPAC Name 2-(2-hydroxyphenoxy)acetic acid
Molecular Weight (g/mol) 168.15
PubChem CID 22783
Formula Weight 168.15
Percent Purity ≥98%
Chemical Name or Material 2-Hydroxyphenoxyacetic acid
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 228-684-8
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only