missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyphenethyl alcohol, 98%

Catalog No. AC182490050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC182490050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC182490050 Supplier Thermo Scientific Chemicals Supplier No. 182490050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7768-28-7 | C8H10O2 | 138.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7768-28-7
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00002890
InChI Key ABFCOJLLBHXNOU-UHFFFAOYSA-N
Synonym 2-hydroxyphenethyl alcohol, 2-2-hydroxyethyl phenol, 2-hydroxyphenylethanol, o-2-hydroxyethyl phenol, benzeneethanol, 2-hydroxy, 2-2-hydroxyphenyl ethanol, o-hydroxyphenethyl alcohol, 2-o-hydroxyphenyl ethanol, 2-hydroxy-benzeneethanol, beta-o-hydroxyphenyl ethanol
PubChem CID 82200
ChEBI CHEBI:64803
IUPAC Name 2-(2-hydroxyethyl)phenol
SMILES OCCC1=CC=CC=C1O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Brown-Yellow to Yellow
Density 1.1590g/mL
Boiling Point 168°C to 169°C (2.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Refractive Index 1.5560 to 1.5600 (20°C, 589nm)
Linear Formula HOC6H4CH2CH2OH
Quantity 5 g
Beilstein 06,906
Specific Gravity 1.159
Formula Weight 138.17
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material 2-Hydroxyphenethyl alcohol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 231-863-3

RUO – Research Use Only