missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyoctanoic acid, 98+%

Catalog No. AAL0231403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0231403 1 g
AAL0231406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0231403 Supplier Thermo Scientific Chemicals Supplier No. L0231403

CAS: 617-73-2 | C8H16O3 | 160.213 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 617-73-2
Molecular Formula C8H16O3
Molecular Weight (g/mol) 160.213
MDL Number MFCD00014410
InChI Key JKRDADVRIYVCCY-UHFFFAOYSA-N
Synonym 2-hydroxycaprylic acid, hydroxyoctanoate, hydroxycaprylic acid, 2-hydroxy caprylic acid, 2-hydroxy-octanoic acid, octanoic acid, hydroxy, alpha-hydroxycaprylic acid, 2-hydroxy-n-octanoic acid, 2-hydroxyoctanoate, alpha-hydroxyoctanoic acid
PubChem CID 94180
ChEBI CHEBI:86543
IUPAC Name 2-hydroxyoctanoic acid
SMILES CCCCCCC(C(=O)O)O

Specifications

Melting Point 66°C to 71°C
Quantity 1 g
Beilstein 1760638
Formula Weight 160.22
Percent Purity ≥98%
Chemical Name or Material 2-Hydroxyoctanoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 210-524-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only