accessibility menu, dialog, popup

UserName

CAS RN 617-73-2

IUPAC Name: 2-hydroxyoctanoic acid
Synonyms: hydroxyoctanoate hydroxyoctanoic acid α-hydroxy-n-caprylate α-hydroxy-n-caprylic acid
Molecular Weight (g/mol): 160.21
Molecular Formula: C8H16O3
InChi Key: JKRDADVRIYVCCY-UHFFFAOYNA-N
SMILES: CCCCCCC(O)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)