missing translation for 'onlineSavingsMsg'
Learn More

2-(Hydroxymethyl)anthracene 98.0+%, TCI America™

Catalog No. p-7130706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H07501G 1 g
H07505G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H07501G Supplier TCI America Supplier No. H07501G

Chemical Identifiers

CAS 22863-82-7
Molecular Formula C15H12O
Molecular Weight (g/mol) 208.26
MDL Number MFCD00142932
InChI Key ZMUUBYLNJMTHBS-UHFFFAOYSA-N
Synonym 2-Anthracenemethanol
PubChem CID 1201478
IUPAC Name anthracen-2-ylmethanol
SMILES C1=CC=C2C=C3C=C(C=CC3=CC2=C1)CO

Specifications

Melting Point 224°C
Color Yellow
Quantity 1 g
Formula Weight 208.26
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Hydroxymethyl)anthracene