missing translation for 'onlineSavingsMsg'
Learn More

2-[(Hydroxymethyl)amino]ethanol 95.0+%, TCI America™

Catalog No. p-7130912
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H11745G 5 g
H117425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H11745G Supplier TCI America Supplier No. H11745G

Chemical Identifiers

CAS 65184-12-5
Molecular Formula C3H9NO2
Molecular Weight (g/mol) 91.11
MDL Number MFCD00128999
InChI Key NWPCFCBFUXXJIE-UHFFFAOYSA-N
Synonym N-(Hydroxymethyl)ethanolamine
PubChem CID 61929
IUPAC Name 2-[(hydroxymethyl)amino]ethan-1-ol
SMILES OCCNCO

Specifications

Color Yellow
Quantity 5 g
UN Number 1760
Formula Weight 91.11
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material 2-[(Hydroxymethyl)amino]ethanol
EINECSNumber (2)-0299
TSCA No