missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxyisobutyric Acid 98.0+%, TCI America™
$46.60 - $426.87
Chemical Identifiers
| CAS | 594-61-6 |
|---|---|
| Molecular Formula | C4H8O3 |
| Molecular Weight (g/mol) | 104.11 |
| MDL Number | MFCD00004459 |
| InChI Key | BWLBGMIXKSTLSX-UHFFFAOYSA-N |
| Synonym | 2-hydroxyisobutyric acid, 2-methyllactic acid, acetonic acid, 2-hydroxy-2-methylpropionic acid, alpha-hydroxyisobutyric acid, hydroxydimethylacetic acid, acetonate, propanoic acid, 2-hydroxy-2-methyl, alpha-hydroxyisobutanoic acid, 2-hydroxyisobutyricacid |
| PubChem CID | 11671 |
| ChEBI | CHEBI:50129 |
| IUPAC Name | 2-hydroxy-2-methylpropanoic acid |
| SMILES | CC(C)(O)C(O)=O |
Chemical Identifiers
| 594-61-6 | |
| 104.11 | |
| BWLBGMIXKSTLSX-UHFFFAOYSA-N | |
| 11671 | |
| 2-hydroxy-2-methylpropanoic acid |
| C4H8O3 | |
| MFCD00004459 | |
| 2-hydroxyisobutyric acid, 2-methyllactic acid, acetonic acid, 2-hydroxy-2-methylpropionic acid, alpha-hydroxyisobutyric acid, hydroxydimethylacetic acid, acetonate, propanoic acid, 2-hydroxy-2-methyl, alpha-hydroxyisobutanoic acid, 2-hydroxyisobutyricacid | |
| CHEBI:50129 | |
| CC(C)(O)C(O)=O |
Specifications
| 594-61-6 | |
| White | |
| MFCD00004459 | |
| 2-hydroxyisobutyric acid, 2-methyllactic acid, acetonic acid, 2-hydroxy-2-methylpropionic acid, alpha-hydroxyisobutyric acid, hydroxydimethylacetic acid, acetonate, propanoic acid, 2-hydroxy-2-methyl, alpha-hydroxyisobutanoic acid, 2-hydroxyisobutyricacid | |
| CC(C)(O)C(O)=O | |
| 104.11 | |
| CHEBI:50129 | |
| ≥98.0% (T) | |
| 2-Hydroxyisobutyric Acid |
| 80°C | |
| C4H8O3 | |
| 25 g | |
| BWLBGMIXKSTLSX-UHFFFAOYSA-N | |
| 2-hydroxy-2-methylpropanoic acid | |
| 11671 | |
| 104.11 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (2)-1429
TSCA : Yes