missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxyisobutyric Acid 98.0+%, TCI America™
Supplier: TCI America M0360100G
Specifications
| 2-Hydroxyisobutyric Acid | |
| 80°C | |
| C4H8O3 | |
| 100 g | |
| BWLBGMIXKSTLSX-UHFFFAOYSA-N | |
| 2-hydroxy-2-methylpropanoic acid | |
| 11671 | |
| 104.11 | |
| Crystalline Powder |
| 594-61-6 | |
| White | |
| MFCD00004459 | |
| 2-hydroxyisobutyric acid, 2-methyllactic acid, acetonic acid, 2-hydroxy-2-methylpropionic acid, alpha-hydroxyisobutyric acid, hydroxydimethylacetic acid, acetonate, propanoic acid, 2-hydroxy-2-methyl, alpha-hydroxyisobutanoic acid, 2-hydroxyisobutyricacid | |
| CC(C)(O)C(O)=O | |
| 104.11 | |
| CHEBI:50129 | |
| ≥98.0% (T) |
Chemical Identifiers
| 594-61-6 | |
| 104.11 | |
| BWLBGMIXKSTLSX-UHFFFAOYSA-N | |
| 11671 | |
| 2-hydroxy-2-methylpropanoic acid |
| C4H8O3 | |
| MFCD00004459 | |
| 2-hydroxyisobutyric acid, 2-methyllactic acid, acetonic acid, 2-hydroxy-2-methylpropionic acid, alpha-hydroxyisobutyric acid, hydroxydimethylacetic acid, acetonate, propanoic acid, 2-hydroxy-2-methyl, alpha-hydroxyisobutanoic acid, 2-hydroxyisobutyricacid | |
| CHEBI:50129 | |
| CC(C)(O)C(O)=O |
Safety and Handling
EINECSNumber : (2)-1429
TSCA : Yes