missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyhippuric acid, 95%

CAS: 487-54-7 | C9H9NO4 | 195.17 g/mol

$643.57 - $643.57

Chemical Identifiers

CAS 487-54-7
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.17
MDL Number MFCD00002695
InChI Key ONJSZLXSECQROL-UHFFFAOYSA-N
Synonym salicyluric acid, 2-hydroxyhippuric acid, salicylurate, o-hydroxyhippuric acid, n-salicyloylglycine, salicyloylglycine, n-2-hydroxybenzoyl glycine, glycine, n-2-hydroxybenzoyl, 2-2-hydroxybenzamido acetic acid, n-o-hydroxybenzoylglycine
PubChem CID 10253
ChEBI CHEBI:9008
IUPAC Name 2-[(2-hydroxybenzoyl)amino]acetic acid
SMILES OC(=O)CNC(=O)C1=CC=CC=C1O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC158620100
SDS View Documents
Thermo Scientific Chemicals
158620100
10 g Glass bottle
Each for $643.57
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

487-54-7
195.17
ONJSZLXSECQROL-UHFFFAOYSA-N
10253
2-[(2-hydroxybenzoyl)amino]acetic acid
C9H9NO4
MFCD00002695
salicyluric acid, 2-hydroxyhippuric acid, salicylurate, o-hydroxyhippuric acid, n-salicyloylglycine, salicyloylglycine, n-2-hydroxybenzoyl glycine, glycine, n-2-hydroxybenzoyl, 2-2-hydroxybenzamido acetic acid, n-o-hydroxybenzoylglycine
CHEBI:9008
OC(=O)CNC(=O)C1=CC=CC=C1O

Specifications

487-54-7
100.0
White to Yellow
94% min. (HPLC)
C9H9NO4
MFCD00002695
10, 92
Solubility in water: soluble. Other solubilities: soluble in ethanol
OC(=O)CNC(=O)C1=CC=CC=C1O
195.17
CHEBI:9008
95%
2-Hydroxyhippuric acid, 97%
96.0
164°C to 169°C
Authentic
Glass bottle
HOC6H4CONHCH2CO2H
10 g
salicyluric acid, 2-hydroxyhippuric acid, salicylurate, o-hydroxyhippuric acid, n-salicyloylglycine, salicyloylglycine, n-2-hydroxybenzoyl glycine, glycine, n-2-hydroxybenzoyl, 2-2-hydroxybenzamido acetic acid, n-o-hydroxybenzoylglycine
ONJSZLXSECQROL-UHFFFAOYSA-N
2-[(2-hydroxybenzoyl)amino]acetic acid
10253
195.17
Powder, Crystals or Chunks
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath

GHS Signal Word: Warning

EINECSNumber : 207-661-6

RTECSNumber : MR8160000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only