missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyhippuric acid, 95%

Catalog No. AC158620100
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC158620100 10 g Glass bottle
AC158620025 2.5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC158620100 Supplier Thermo Scientific Chemicals Supplier No. 158620100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 487-54-7 | C9H9NO4 | 195.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 487-54-7
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.17
MDL Number MFCD00002695
InChI Key ONJSZLXSECQROL-UHFFFAOYSA-N
Synonym salicyluric acid, 2-hydroxyhippuric acid, salicylurate, o-hydroxyhippuric acid, n-salicyloylglycine, salicyloylglycine, n-2-hydroxybenzoyl glycine, glycine, n-2-hydroxybenzoyl, 2-2-hydroxybenzamido acetic acid, n-o-hydroxybenzoylglycine
PubChem CID 10253
ChEBI CHEBI:9008
IUPAC Name 2-[(2-hydroxybenzoyl)amino]acetic acid
SMILES OC(=O)CNC(=O)C1=CC=CC=C1O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 164°C to 169°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Linear Formula HOC6H4CONHCH2CO2H
Quantity 10 g
Beilstein 10, 92
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
Formula Weight 195.17
Percent Purity 95%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material 2-Hydroxyhippuric acid, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath
EINECSNumber 207-661-6
RTECSNumber MR8160000
TSCA TSCA

RUO – Research Use Only